Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SORBS1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-06079
103409-166
Anti-SORBS1 Goat Polyclonal Antibody
The SORBS1 Antibody from Novus Biologicals is a goat polyclonal antibody to SORBS1. This antibody reacts with human, mouse. The SORBS1 Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: SORBS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-SERNDWEPPDKKVD
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty