Anti-SODD Mouse Polyclonal Antibody

H00009530-B01P
103337-166EA 583.08 USD
103337-166
Anti-SODD Mouse Polyclonal Antibody
The SODD / BAG4 Antibody from Novus Biologicals is a mouse polyclonal antibody to SODD / BAG4. This antibody reacts with human. The SODD / BAG4 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SODD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR