Anti-SODD Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56696
103361-168EA 417.23 USD
103361-168
Anti-SODD Rabbit Polyclonal Antibody
Antibodies
The SODD / BAG4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SODD / BAG4. This antibody reacts with human. The SODD / BAG4 Antibody has been validated for the following applications: Western Blot, Flow Cytometry.

Type: Primary
Antigen: SODD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR