Anti-SOD2 Rabbit Polyclonal Antibody

H00006648-D01P
103310-504EA 583.08 USD
103310-504
Anti-SOD2 Rabbit Polyclonal Antibody
The SOD2 / Mn-SOD Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOD2 / Mn-SOD. This antibody reacts with human. The SOD2 / Mn-SOD Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR