Anti-SOD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20534
102711-938EA 635.51 USD
102711-938
Anti-SOD1 Rabbit Polyclonal Antibody
Antibodies
The SOD1 / Cu-Zn SOD Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOD1 / Cu-Zn SOD. This antibody reacts with human, mouse, rat. The SOD1 / Cu-Zn SOD Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR