Anti-SOD1 Mouse Monoclonal Antibody [clone: 10D5]

H00006647-M04
103310-500EA 583.08 USD
103310-500
Anti-SOD1 Mouse Monoclonal Antibody [clone: 10D5]
The SOD1 / Cu-Zn SOD Antibody (10D5) from Novus Biologicals is a mouse monoclonal antibody to SOD1 / Cu-Zn SOD. This antibody reacts with human. The SOD1 / Cu-Zn SOD Antibody (10D5) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SOD1
Clonality: Monoclonal
Clone: 10D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR