Anti-SOCS-5 Mouse Polyclonal Antibody

H00009655-B01P
103337-440EA 583.08 USD
103337-440
Anti-SOCS-5 Mouse Polyclonal Antibody
The SOCS-5 Antibody from Novus Biologicals is a mouse polyclonal antibody to SOCS-5. This antibody reacts with human. The SOCS-5 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SOCS-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR