Anti-SOCS-4 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00122809-B01P
103346-528EA 399 USD
103346-528
Anti-SOCS-4 Mouse Polyclonal Antibody
Antibodies
The SOCS-4 Antibody from Novus Biologicals is a mouse polyclonal antibody to SOCS-4. This antibody reacts with human. The SOCS-4 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SOCS-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR