Anti-SOCS-3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20451
102711-772EA 379 USD
102711-772
Anti-SOCS-3 Rabbit Polyclonal Antibody
The SOCS-3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOCS-3. This antibody reacts with human, mouse. The SOCS-3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SOCS-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR