Anti-SOCS-3 Mouse Polyclonal Antibody

H00009021-B01P
103336-058EA 553.85 USD
103336-058
Anti-SOCS-3 Mouse Polyclonal Antibody
The SOCS-3 Antibody from Novus Biologicals is a mouse polyclonal antibody to SOCS-3. This antibody reacts with human. The SOCS-3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SOCS-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR