Anti-SOCS-3 Mouse Monoclonal Antibody [clone: 1F6]

H00009021-M03
103336-064EA 553.85 USD
103336-064
Anti-SOCS-3 Mouse Monoclonal Antibody [clone: 1F6]
The SOCS-3 Antibody (1F6) from Novus Biologicals is a mouse monoclonal antibody to SOCS-3. This antibody reacts with human. The SOCS-3 Antibody (1F6) has been validated for the following applications: ELISA.

Type: Primary
Antigen: SOCS-3
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR