Anti-SOCS-2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
29920002-0.1MG
103317-102EA 349 USD
103317-102
Anti-SOCS-2 Rabbit Polyclonal Antibody
Antibodies
The SOCS-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOCS-2. This antibody reacts with human. The SOCS-2 Antibody has been validated for the following applications: ELISA.

Type: Primary
Antigen: SOCS-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR