Anti-SNX29 Mouse Monoclonal Antibody [clone: 6D6]

Supplier: Novus Biologicals
H00084127-M02
103345-064EA 798.46 USD
103345-064
Anti-SNX29 Mouse Monoclonal Antibody [clone: 6D6]
Antibodies
The SNX29 Antibody (6D6) from Novus Biologicals is a mouse monoclonal antibody to SNX29. This antibody reacts with human, mouse. The SNX29 Antibody (6D6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SNX29
Clonality: Monoclonal
Clone: 6D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR