Anti-SNX25 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00083891-B01P
103344-966EA 798.46 USD
103344-966
Anti-SNX25 Mouse Polyclonal Antibody
Antibodies
The SNX25 Antibody from Novus Biologicals is a mouse polyclonal antibody to SNX25. This antibody reacts with human. The SNX25 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SNX25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR