Anti-SNX24 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00028966-B01P
103342-908EA 798.46 USD
103342-908
Anti-SNX24 Mouse Polyclonal Antibody
Antibodies
The SNX24 Antibody from Novus Biologicals is a mouse polyclonal antibody to SNX24. This antibody reacts with human. The SNX24 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SNX24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR