Anti-SNX24 Mouse Monoclonal Antibody [clone: 2A11]

Supplier: Novus Biologicals
H00028966-M01
103342-910EA 798.46 USD
103342-910
Anti-SNX24 Mouse Monoclonal Antibody [clone: 2A11]
Antibodies
The SNX24 Antibody (2A11) from Novus Biologicals is a mouse monoclonal antibody to SNX24. This antibody reacts with human. The SNX24 Antibody (2A11) has been validated for the following applications: ELISA.

Type: Primary
Antigen: SNX24
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR