Anti-SNX16 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-85974
103272-368EA 734.58 USD
103272-368
Anti-SNX16 Rabbit Polyclonal Antibody
Antibodies
The SNX16 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNX16. This antibody reacts with human. The SNX16 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SNX16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR