Anti-SNX15 Mouse Monoclonal Antibody [clone: 1D4]

Supplier: Novus Biologicals
H00029907-M01
103343-118EA 399 USD
103343-118
Anti-SNX15 Mouse Monoclonal Antibody [clone: 1D4]
Antibodies
The SNX15 Antibody (1D4) from Novus Biologicals is a mouse monoclonal antibody to SNX15. This antibody reacts with human. The SNX15 Antibody (1D4) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SNX15
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR