Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SNX10 Mouse Monoclonal Antibody [clone: 1G5]

Supplier: Novus Biologicals
H00029887-M01
103343-102
Anti-SNX10 Mouse Monoclonal Antibody [clone: 1G5]
The SNX10 Antibody (1G5) from Novus Biologicals is a mouse monoclonal antibody to SNX10. This antibody reacts with human. The SNX10 Antibody (1G5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SNX10
Clonality: Monoclonal
Clone: 1G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty