Anti-SNX1 Mouse Monoclonal Antibody [clone: 6H1]

Supplier: Novus Biologicals
H00006642-M01
103310-490EA 798.46 USD
103310-490
Anti-SNX1 Mouse Monoclonal Antibody [clone: 6H1]
Antibodies
The SNX1 Antibody (6H1) from Novus Biologicals is a mouse monoclonal antibody to SNX1. This antibody reacts with human. The SNX1 Antibody (6H1) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin, RNA Inhibition.

Type: Primary
Antigen: SNX1
Clonality: Monoclonal
Clone: 6H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR