Anti-SnoN Mouse Monoclonal Antibody [clone: 1C12]

H00006498-M01
103310-192EA 583.08 USD
103310-192
Anti-SnoN Mouse Monoclonal Antibody [clone: 1C12]
The SnoN Antibody (1C12) from Novus Biologicals is a mouse monoclonal antibody to SnoN. This antibody reacts with human. The SnoN Antibody (1C12) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SnoN
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR