Anti-SNF5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00006598-D03P
103310-326EA 429 USD
103310-326
Anti-SNF5 Rabbit Polyclonal Antibody
Antibodies
The SNF5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNF5. This antibody reacts with human. The SNF5 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SNF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR