Anti-SNF1LK2/SIK2 Mouse Monoclonal Antibody [clone: 4C6]

Supplier: Novus Biologicals
H00023235-M03
103341-132EA 798.46 USD
103341-132
Anti-SNF1LK2/SIK2 Mouse Monoclonal Antibody [clone: 4C6]
Antibodies
The SNF1LK2 / SIK2 Antibody (4C6) from Novus Biologicals is a mouse monoclonal antibody to SNF1LK2 / SIK2. This antibody reacts with human. The SNF1LK2 / SIK2 Antibody (4C6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SNF1LK2/SIK2
Clonality: Monoclonal
Clone: 4C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR