Anti-SNF1LK2/SIK2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56458SS
103359-170EA 208 USD
103359-170
Anti-SNF1LK2/SIK2 Rabbit Polyclonal Antibody
Antibodies
The SNF1LK2 / SIK2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNF1LK2 / SIK2. This antibody reacts with human, mouse, rat, bovine, canine, primate. The SNF1LK2 / SIK2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SNF1LK2/SIK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR