Anti-SNAPC5 Mouse Monoclonal Antibody [clone: 500]

Supplier: Novus Biologicals
H00010302-M03
103338-630EA 798.46 USD
103338-630
Anti-SNAPC5 Mouse Monoclonal Antibody [clone: 500]
Antibodies
The SNAPC5 Antibody (5E2) from Novus Biologicals is a mouse monoclonal antibody to SNAPC5. This antibody reacts with human. The SNAPC5 Antibody (5E2) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SNAPC5
Clonality: Monoclonal
Clone: 500
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR