Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SNAPC5 Mouse Monoclonal Antibody [clone: 6G10]

Supplier: Novus Biologicals
H00010302-M07
103338-632
Anti-SNAPC5 Mouse Monoclonal Antibody [clone: 6G10]
The SNAPC5 Antibody (6G10) from Novus Biologicals is a mouse monoclonal antibody to SNAPC5. This antibody reacts with human. The SNAPC5 Antibody (6G10) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SNAPC5
Clonality: Monoclonal
Clone: 6G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty