Anti-SNAPC4 Mouse Monoclonal Antibody [clone: 1D1]

Supplier: Novus Biologicals
H00006621-M01
103310-440EA 798.46 USD
103310-440
Anti-SNAPC4 Mouse Monoclonal Antibody [clone: 1D1]
Antibodies
The SNAPC4 Antibody (1D1) from Novus Biologicals is a mouse monoclonal antibody to SNAPC4. This antibody reacts with human. The SNAPC4 Antibody (1D1) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SNAPC4
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR