Anti-SNAP25 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00006616-D01P
103310-426EA 429 USD
103310-426
Anti-SNAP25 Rabbit Polyclonal Antibody
Antibodies
The SNAP25 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNAP25. This antibody reacts with human. The SNAP25 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SNAP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR