Anti-Snail Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00006615-B02P
103310-416EA 399 USD
103310-416
Anti-Snail Mouse Polyclonal Antibody
Antibodies
The Snail Antibody from Novus Biologicals is a mouse polyclonal antibody to Snail. This antibody reacts with human. The Snail Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Snail
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR