Anti-SMYD3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20430
102711-730EA 635.51 USD
102711-730
Anti-SMYD3 Rabbit Polyclonal Antibody
Antibodies
The SMYD3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMYD3. This antibody reacts with human, mouse, rat. The SMYD3 Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: SMYD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR