Anti-SmyD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20428
102711-726EA 635.51 USD
102711-726
Anti-SmyD1 Rabbit Polyclonal Antibody
Antibodies
The SmyD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SmyD1. This antibody reacts with human, mouse, rat. The SmyD1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SmyD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR