Anti-SMPDL3B Mouse Monoclonal Antibody [clone: 5E12]

Supplier: Novus Biologicals
H00027293-M01
103342-756EA 798.46 USD
103342-756
Anti-SMPDL3B Mouse Monoclonal Antibody [clone: 5E12]
Antibodies
The SMPDL3B Antibody (5E12) from Novus Biologicals is a mouse monoclonal antibody to SMPDL3B. This antibody reacts with human. The SMPDL3B Antibody (5E12) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SMPDL3B
Clonality: Monoclonal
Clone: 5E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR