Anti-SMPDL3A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88862
103276-140EA 734.58 USD
103276-140
Anti-SMPDL3A Rabbit Polyclonal Antibody
Antibodies
The SMPDL3A Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMPDL3A. This antibody reacts with human. The SMPDL3A Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SMPDL3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR