Anti-SMPD1 Mouse Monoclonal Antibody [clone: 4H2]

Supplier: Novus Biologicals
H00006609-M01
103310-394EA 798.46 USD
103310-394
Anti-SMPD1 Mouse Monoclonal Antibody [clone: 4H2]
Antibodies
The SMPD1 Antibody (4H2) from Novus Biologicals is a mouse monoclonal antibody to SMPD1. This antibody reacts with human. The SMPD1 Antibody (4H2) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SMPD1
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR