Anti-SMOC-2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20425
102711-720EA 635.51 USD
102711-720
Anti-SMOC-2 Rabbit Polyclonal Antibody
Antibodies
The SMOC-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMOC-2. This antibody reacts with human. The SMOC-2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SMOC-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR