Anti-SMN Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2B1]

Supplier: Novus Biologicals
NB100-1936F
103368-778EA 675.69 USD
103368-778
Anti-SMN Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2B1]
Antibodies
The SMN Antibody (2B1) [FITC] from Novus Biologicals is a mouse monoclonal antibody to SMN. This antibody reacts with human, mouse, rat, primate, xenopus. The SMN Antibody (2B1) [FITC] has been validated for the following applications: ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SMN
Clonality: Monoclonal
Clone: 2B1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primate, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR