Anti-SMG9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13355
103283-692EA 734.58 USD
103283-692
Anti-SMG9 Rabbit Polyclonal Antibody
Antibodies
The SMG9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMG9. This antibody reacts with human. The SMG9 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SMG9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR