Anti-SMG6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-92410
103284-290EA 734.58 USD
103284-290
Anti-SMG6 Rabbit Polyclonal Antibody
Antibodies
The SMG6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMG6. This antibody reacts with human. The SMG6 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).

Type: Primary
Antigen: SMG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR