Anti-SMG1 Mouse Monoclonal Antibody [clone: 2B10]

Supplier: Novus Biologicals
H00023049-M04
103340-938EA 798.46 USD
103340-938
Anti-SMG1 Mouse Monoclonal Antibody [clone: 2B10]
Antibodies
The SMG1 Antibody (2B10) from Novus Biologicals is a mouse monoclonal antibody to SMG1. This antibody reacts with human. The SMG1 Antibody (2B10) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SMG1
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR