Anti-SMCR8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-93834
103283-484EA 734.58 USD
103283-484
Anti-SMCR8 Rabbit Polyclonal Antibody
Antibodies
The SMCR8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMCR8. This antibody reacts with human. The SMCR8 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SMCR8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR