Anti-SMCHD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-82978
103266-344EA 777.05 USD
103266-344
Anti-SMCHD1 Rabbit Polyclonal Antibody
Antibodies
The SMCHD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMCHD1. This antibody reacts with human. The SMCHD1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SMCHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR