Anti-SMC5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
40310002-0.1MG
103317-442EA 549 USD
103317-442
Anti-SMC5 Rabbit Polyclonal Antibody
Antibodies
The SMC5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SMC5. This antibody reacts with drosophila. The SMC5 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SMC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR