Anti-SMC5 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46115
103409-164EA 329 USD
103409-164
Anti-SMC5 Goat Polyclonal Antibody
Antibodies
The SMC5 Antibody from Novus Biologicals is a goat polyclonal antibody to SMC5. This antibody reacts with human, mouse, rat, bovine, canine. The SMC5 Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: SMC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal region
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR