Anti-SMC1L2 Mouse Monoclonal Antibody [clone: 6A10]

Supplier: Novus Biologicals
H00027127-M01
103342-596EA 798.46 USD
103342-596
Anti-SMC1L2 Mouse Monoclonal Antibody [clone: 6A10]
Antibodies
The SMC1L2 Antibody (6A10) from Novus Biologicals is a mouse monoclonal antibody to SMC1L2. This antibody reacts with human. The SMC1L2 Antibody (6A10) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SMC1L2
Clonality: Monoclonal
Clone: 6A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR