Anti-S100A9 Mouse Monoclonal Antibody [clone: 4G9]

H00006280-M11
103309-730EA 553.85 USD
103309-730
Anti-S100A9 Mouse Monoclonal Antibody [clone: 4G9]
The S100A9 Antibody (4G9) from Novus Biologicals is a mouse monoclonal antibody to S100A9. This antibody reacts with human. The S100A9 Antibody (4G9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone: 4G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR