Anti-RSK3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00006196-D01P
103309-536EA 429 USD
103309-536
Anti-RSK3 Rabbit Polyclonal Antibody
Antibodies
The RSK3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RSK3. This antibody reacts with human. The RSK3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: RSK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR