Anti-RRM2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00006241-D01P
103309-620EA 429 USD
103309-620
Anti-RRM2 Rabbit Polyclonal Antibody
Antibodies
The RRM2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RRM2. This antibody reacts with human. The RRM2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: RRM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR