Anti-RPL28 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13254
103285-630EA 734.58 USD
103285-630
Anti-RPL28 Rabbit Polyclonal Antibody
Antibodies
The RPL28 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RPL28. This antibody reacts with human. The RPL28 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RPL28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR