Anti-RPL23A Mouse Monoclonal Antibody [clone: 3E11.]

Supplier: Novus Biologicals
H00006147-M10
103309-474EA 798.46 USD
103309-474
Anti-RPL23A Mouse Monoclonal Antibody [clone: 3E11.]
Antibodies
The RPL23A Antibody (3E11.) from Novus Biologicals is a mouse monoclonal antibody to RPL23A. This antibody reacts with human. The RPL23A Antibody (3E11.) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: RPL23A
Clonality: Monoclonal
Clone: 3E11.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR