Anti-PPAN Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88525
103274-918EA 734.58 USD
103274-918
Anti-PPAN Rabbit Polyclonal Antibody
Antibodies
The PPAN Antibody from Novus Biologicals is a rabbit polyclonal antibody to PPAN. This antibody reacts with human. The PPAN Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PPAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR